Learn More
Fisherbrand™ Test Tubes, Soda lime, Without Rim, Medium Wall
Test tubes with round bottom are universal cylindrical laboratory glassware that are used e.g. for chemical reactions, pretreatments or for storing small amounts of liquid.
Brand:  Fisherbrand™ 90101214
 View more versions of this product
Product Code. 17921122
  • 132.00 € / Pack of 1000
Explore more special offers
Description
Description
  • Transferring substances and liquids into other states of aggregation and thus initiating further reactions is very important for the daily work in the laboratory. A wide variety of vessels, apparatus and equipment are used for these work. Reaction controls, test series and preliminary samples are usually carried out in test tubes due to the small amounts of substance removed.
  • Manufactured from soda lime glass
  • Thick-walled and therefore mechanically very resistant
  • Available with beaded rim or straight rim
  • With round bottom
TRUSTED_SUSTAINABILITY
Specifications
Specifications
12 mL
Soda Lime Glass
Non-sterile
No
Not Included
0.9 mm
100 mm
Cylindrical
Not Autoclavable
16 mm
Test Tube
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products