missing translation for 'onlineSavingsMsg'
Learn More

Tetra-N-Butyl Ammonium Hydroxide, for HPLC, Solution 0.5M,Certified, Fisher Chemical

Product Code. 10366150
Click to view available options
Quantity:
250 mL
Packaging:
HDPE plastic bottle

Specifications

CAS 2052-49-5
Color Colorless
pH 12
Physical Form Liquid
Quantity 250 mL
Molecular Formula C16H37NO
MDL Number MFCD00009425
Synonym tetrabutylammonium hydroxide, tetra-n-butylammonium hydroxide, tetrabutylazanium hydroxide, tetrabutylammoniumhydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide, ammonium, tetrabutyl-, hydroxide, n,n,n-tributyl-1-butanaminium hydroxide, tetra n-butyl ammonium hydroxide, tetra-n-butyl ammonium hydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide 1:1
InChI Key VDZOOKBUILJEDG-UHFFFAOYSA-M
SMILES [OH-].CCCC[N+](CCCC)(CCCC)CCCC
IUPAC Name tetrabutylazanium hydroxide
Molecular Weight (g/mol) 259.48
PubChem CID 2723671
Formula Weight 259.47g/mol
Packaging HDPE plastic bottle
Show More Show Less
compliance-icons
Product Identifier
  • Tetrabutylammonium hydroxide, solution 0.5M
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : 1-Butanaminium, N, N, N-tributyl-, hydroxide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.