missing translation for 'onlineSavingsMsg'
Learn More
Tetra-n-butylammonium chloride hydrate, Thermo Scientific Chemicals Product Code.: 11363527

Tetra-n-butylammonium chloride hydrate, Thermo Scientific Chemicals

Product Code. 11363527
5 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11363527

Brand: Thermo Scientific Alfa Aesar 019347.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Tetra-n-butylammonium chloride hydrate is used with phosphorus pentoxide for greener deoxychlorination. It is used as a phase transfer catalyst for the preparation of 2-(3-pyridyl)-1-methylpyrrole by the reaction between 3-bromopyridine and 1-methyl-2pyrrolecarboxylic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37451-68-6
Molecular Formula C16H36ClN
Molecular Weight (g/mol) 277.92
MDL Number MFCD00011635
InChI Key NHGXDBSUJJNIRV-UHFFFAOYSA-M
Synonym tetrabutylammonium chloride hydrate, tetrabutyl ammonium chloride hydrate, tetrabutylammonium chloride monohydrate, tetra-n-butylammonium chloride hydrate, tetrabutylammoniumchlorideh2o, 1-butanaminium, n,n,n-tributyl-, chloride, monohydrate, tbac hydrate, bu4ncl.h2o, n-bu4ncl.h2o, ksc222g0b
PubChem CID 16212273
SMILES [Cl-].CCCC[N+](CCCC)(CCCC)CCCC

Specifications

Melting Point 73°C to 75°C
Quantity 5 g
Sensitivity Hygroscopic
Solubility Information Soluble in cold water.
IUPAC Name tetrabutylazanium chloride
Formula Weight 277.92 (Anhydrous)
Physical Form Crystalline
Chemical Name or Material Tetra-n-butylammonium chloride hydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tetra-n-butylammonium chloride hydrate, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.