Learn More
Tetra-n-butylammonium chloride hydrate, Thermo Scientific Chemicals
Description
Tetra-n-butylammonium chloride hydrate is used with phosphorus pentoxide for greener deoxychlorination. It is used as a phase transfer catalyst for the preparation of 2-(3-pyridyl)-1-methylpyrrole by the reaction between 3-bromopyridine and 1-methyl-2pyrrolecarboxylic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 73°C to 75°C |
Quantity | 25 g |
Sensitivity | Hygroscopic |
Solubility Information | Soluble in cold water. |
IUPAC Name | tetrabutylazanium chloride |
Formula Weight | 277.92 (Anhydrous) |
Physical Form | Crystalline |
Chemical Name or Material | Tetra-n-butylammonium chloride hydrate |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
C30S034 | Certificate of Analysis | 07/12/2021 | 019347.06,019347.14,019347.22 |
H01Y017 | Certificate of Analysis | 07/12/2021 | 019347.06,019347.14,019347.22 |
T15B051 | Certificate of Analysis | 07/12/2021 | 019347.06,019347.14,019347.22 |
B01M28 | Certificate of Analysis | 07/12/2021 | 019347.06,019347.14,019347.22 |
L01P01 | Certificate of Analysis | 07/12/2021 | 019347.06,019347.14,019347.22 |
Safety and Handling

- Tetra-n-butylammonium chloride hydrate
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- MIXTURE LIST-Contains : Tetrabutylammonium chloride hydrate
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.