missing translation for 'onlineSavingsMsg'
Learn More

Tetra-n-butylammonium phosphate, 99+%, Thermo Scientific Chemicals

Product Code. 11353517
Click to view available options
Quantity:
0.5 g
2 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Used as a counter-ion in ion-pair chromatography of water-soluble vitamins

Tetra-n-butylammonium phosphate is used as a counter-ion in ion-pair reagent in high-performance liquid chromatography for the simultaneous determination of chlorpheniramine and maleate of water-soluble vitamins. It acts as a phase transfer catalyst. Further, it is used as a buffering reagent in HPLC. In addition to this, it is used as a mobile phase additive in two-dimensional reversed-phase high-performance thin-layer chromatography for the separation of chenodeoxycholic acid and deoxycholic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5574-97-0
Molecular Formula C16H38NO4P
Molecular Weight (g/mol) 339.46
MDL Number MFCD00064526
InChI Key ARRNBPCNZJXHRJ-UHFFFAOYSA-M
Synonym tetrabutylammonium dihydrogen phosphate, tetrabutylammonium phosphate, tetrabutylammonium dihydrogenphosphate, 1-butanaminium, n,n,n-tributyl-, phosphate 1:1, tetra-n-butylammonium phosphate, tetrabutylazanium dihydrogen phosphate, dihydrogen phosphate; tetrabutylazanium, dihydrogen phosphate; tetrabutylammonium, tetrabutyl ammonium dihydrogen phosphate, tetrabutylammonium ion dihydrogenphosphate ion
PubChem CID 2735142
IUPAC Name dihydrogen phosphate;tetrabutylazanium
SMILES OP(O)([O-])=O.CCCC[N+](CCCC)(CCCC)CCCC

Specifications

Melting Point 151°C to 154°C
Color White
Linear Formula [CH3(CH2)3]4NH2PO4
Quantity 0.5 g
Beilstein 5196532
Solubility Information Soluble in acetonitrile,ethanol and water.
Formula Weight 339.46
Percent Purity ≥99%
Physical Form Crystalline
Chemical Name or Material Tetra-n-butylammonium phosphate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.