missing translation for 'onlineSavingsMsg'
Learn More

Tetra-n-butyllead, 95%, Thermo Scientific™

Product Code. 11428680
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1920-90-7
Molecular Formula C16H36Pb
Molecular Weight (g/mol) 435.664
MDL Number MFCD00015226
InChI Key KDQHJGWPOQNCMI-UHFFFAOYSA-N
Synonym plumbane, tetrabutyl, tetra-n-butyllead, lead tetra-n-butyl, tetra-n-butyl lead, lead tetrabutyl, phthalocyanine, ferric, kdqhjgwpoqncmi-uhfffaoysa
PubChem CID 15978
IUPAC Name tetrabutylplumbane
SMILES CCCC[Pb](CCCC)(CCCC)CCCC

Specifications

Color Colorless to Yellow
Density 1.324 g/mL
Boiling Point 140°C (1 mmHg)
Flash Point 140°C (284°F)
Linear Formula [CH3(CH2)3]4Pb
Refractive Index 1.5119
Quantity 1 g
UN Number UN3282
Sensitivity Air and light sensitive
Solubility Information Insoluble in water
Formula Weight 435.66
Percent Purity 95%
Physical Form Liquid, May Include Precipitate
Chemical Name or Material Tetra-n-butyllead
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'