missing translation for 'onlineSavingsMsg'
Learn More

Tetra-n-hexylammonium benzoate, ca 85%, water ca 10%, Thermo Scientific™

Product Code. 11430878
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16436-29-6
Molecular Formula C31H57NO2
Molecular Weight (g/mol) 475.80
MDL Number MFCD00041982
InChI Key PSEQWFPWQRZBOO-UHFFFAOYSA-M
Synonym tetrahexylammonium benzoate, 1-hexanaminium, n,n,n-trihexyl-, benzoate, tetrahexylazanium benzoate, tetra-n-hexylammonium benzoate, 1-hexanaminium, n,n,n-trihexyl-, benzoate 1:1, tetrahexylazanium, thab, ammonium, tetrahexyl-, benzoate, ammonium, benzoate, tetrahexylazanium,benzoate
PubChem CID 85419
IUPAC Name tetrahexylazanium;benzoate
SMILES [O-]C(=O)C1=CC=CC=C1.CCCCCC[N+](CCCCCC)(CCCCCC)CCCCCC

Specifications

Quantity 1 g
Formula Weight 456.73
Percent Purity ∼85%
Assay water ≈10%
Chemical Name or Material Tetra-n-hexylammonium benzoate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'