missing translation for 'onlineSavingsMsg'
Learn More

Tetra-n-hexylammonium perchlorate, Thermo Scientific Chemicals

Product Code. 11307368
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4656-81-9
Molecular Formula C24H52ClNO4
Molecular Weight (g/mol) 454.13
MDL Number MFCD00043185
InChI Key RLCKPGXHGKSGOS-UHFFFAOYSA-M
Synonym tetra-n-hexylammonium perchlorate, tetrahexylammonium perchlorate, tetrahexylazanium perchlorate, 1-hexanaminium, n,n,n-trihexyl-, perchlorate, n,n,n-trihexylhexan-1-aminiumperchlorate, 1-hexanaminium, n,n,n-trihexyl-, perchlorate 1:1, tetra-n-hexyl ammonium perchlorate
PubChem CID 78388
SMILES [O-][Cl](=O)(=O)=O.CCCCCC[N+](CCCCCC)(CCCCCC)CCCCCC

Specifications

Quantity 5 g
UN Number UN1479
IUPAC Name tetrahexylazanium;perchlorate
Formula Weight 454.13
Physical Form Crystalline
Chemical Name or Material Tetra-n-hexylammonium perchlorate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'