Learn More
Tetraallyltin, 96%, Thermo Scientific Chemicals
101.51 € - 355.77 €
Chemical Identifiers
CAS | 7393-43-3 |
---|---|
Molecular Formula | C12H20Sn |
Molecular Weight (g/mol) | 283.00 |
MDL Number | MFCD00008637 |
InChI Key | XJPKDRJZNZMJQM-UHFFFAOYSA-N |
Synonym | tetraallyltin, tetraallylstannane, tetrallylstannane, stannane, tetra-2-propenyl, stannane, tetraallyl, acmc-209osy, tetrakis prop-2-enyl stannane, stannane,tetra-2-propen-1-yl, tetrakis prop-2-en-1-yl stannane |
PubChem CID | 81878 |
IUPAC Name | tetrakis(prop-2-enyl)stannane |
SMILES | C=CC[Sn](CC=C)(CC=C)CC=C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11374849
|
Thermo Scientific Alfa Aesar
071158.04 |
2 g |
101.51 €
2g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11384849
|
Thermo Scientific Alfa Aesar
071158.09 |
10 g |
355.77 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Applied as allyl transfer reagent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7393-43-3 | |
283.00 | |
XJPKDRJZNZMJQM-UHFFFAOYSA-N | |
81878 | |
C=CC[Sn](CC=C)(CC=C)CC=C |
C12H20Sn | |
MFCD00008637 | |
tetraallyltin, tetraallylstannane, tetrallylstannane, stannane, tetra-2-propenyl, stannane, tetraallyl, acmc-209osy, tetrakis prop-2-enyl stannane, stannane,tetra-2-propen-1-yl, tetrakis prop-2-en-1-yl stannane | |
tetrakis(prop-2-enyl)stannane |
Specifications
7393-43-3 | |
69°C to 70°C (1.5 mmHg) | |
C12H20Sn | |
MFCD00008637 | |
Air Sensitive | |
Not miscible in water or difficult to mix. | |
C=CC[Sn](CC=C)(CC=C)CC=C | |
283.00 | |
282.98 | |
Liquid |
1.179 g/mL | |
75°C (167°F) | |
1.5385 | |
UN2788 | |
tetraallyltin, tetraallylstannane, tetrallylstannane, stannane, tetra-2-propenyl, stannane, tetraallyl, acmc-209osy, tetrakis prop-2-enyl stannane, stannane,tetra-2-propen-1-yl, tetrakis prop-2-en-1-yl stannane | |
XJPKDRJZNZMJQM-UHFFFAOYSA-N | |
tetrakis(prop-2-enyl)stannane | |
81878 | |
96% | |
Tetraallyltin |
Safety and Handling
GHS H Statement
H300-H310-H330-H227-H315-H319
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
P210-P235-P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P330-P361-P363-P370+P378q-P501c
H227-H301+H311+H331-H335
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.
missing translation for 'einecsNumber' : 230-987-5
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.