missing translation for 'onlineSavingsMsg'
Learn More

Tetraamminepalladium(II) bromide, Premion™, 99.95% (metals basis), Pd 31.3% min, Thermo Scientific™

Product Code. 11339427
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 44463-62-9
Molecular Formula Br2H12N4Pd
Molecular Weight (g/mol) 334.352
MDL Number MFCD00797352
InChI Key BLPSMPASLSGGOC-UHFFFAOYSA-L
Synonym tetraamminepalladium ii bromide, dibromopalladium tetraamine, dibromopalladium-ammonia 1/4, tetraamminepalladium ii bromide, premion, tetraamminepalladium ii bromide trace metals basis
PubChem CID 16213236
IUPAC Name azane;dibromopalladium
SMILES N.N.N.N.Br[Pd]Br

Specifications

Melting Point >150°C (dec.)
Physical Form Powder
Quantity 1 g
Formula Weight 334.34
Percent Purity 99.95%
Grade Premion™
Concentration or Composition (by Analyte or Components) Pd 31.3% min.
Assay (metals basis)
Chemical Name or Material Tetraamminepalladium(II) bromide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.