missing translation for 'onlineSavingsMsg'
Learn More

Tetraamminepalladium(II) bromide, Premion™, 99.95% (metals basis), Pd 31.3% min, Thermo Scientific™

Chemical Identifiers

CAS 44463-62-9
Molecular Formula Br2H12N4Pd
Molecular Weight (g/mol) 334.352
MDL Number MFCD00797352
InChI Key BLPSMPASLSGGOC-UHFFFAOYSA-L
Synonym tetraamminepalladium ii bromide, dibromopalladium tetraamine, dibromopalladium-ammonia 1/4, tetraamminepalladium ii bromide, premion, tetraamminepalladium ii bromide trace metals basis
PubChem CID 16213236
IUPAC Name azane;dibromopalladium
SMILES N.N.N.N.Br[Pd]Br
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11339427
View Documents Promotion Details
Thermo Scientific Alfa Aesar
041021.03
1 g N/A
N/A
11349427
View Documents Promotion Details
Thermo Scientific Alfa Aesar
041021.06
5 g N/A
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

44463-62-9
334.352
BLPSMPASLSGGOC-UHFFFAOYSA-L
16213236
N.N.N.N.Br[Pd]Br
Br2H12N4Pd
MFCD00797352
tetraamminepalladium ii bromide, dibromopalladium tetraamine, dibromopalladium-ammonia 1/4, tetraamminepalladium ii bromide, premion, tetraamminepalladium ii bromide trace metals basis
azane;dibromopalladium

Specifications

>150°C (dec.)
Powder
MFCD00797352
BLPSMPASLSGGOC-UHFFFAOYSA-L
azane;dibromopalladium
16213236
99.95%
Pd 31.3% min.
Tetraamminepalladium(II) bromide
44463-62-9
Br2H12N4Pd
tetraamminepalladium ii bromide, dibromopalladium tetraamine, dibromopalladium-ammonia 1/4, tetraamminepalladium ii bromide, premion, tetraamminepalladium ii bromide trace metals basis
N.N.N.N.Br[Pd]Br
334.352
334.34
Premion™
(metals basis)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications