missing translation for 'onlineSavingsMsg'
Learn More

Tetraammineplatinum(II) hydrogen phosphate solution, Pt 2% w/w (cont. Pt), Thermo Scientific™

Product Code. 11470660
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 127733-98-6
Molecular Formula H13N4O4PPt
Molecular Weight (g/mol) 359.19
MDL Number MFCD03788258
InChI Key LSDWMQXWIQMHNQ-UHFFFAOYSA-L
Synonym azane;hydrogen phosphate;platinum 2+, platinum 2+ tetraamine hydrogen phosphate, platinum 2+ hydrogen phosphate-ammonia 1/1/4, platinum 2+ ion tetraamine hydrogen phosphate
IUPAC Name platinum(2+) tetraamine hydrogen phosphate
SMILES N.N.N.N.[Pt++].OP([O-])([O-])=O

Specifications

Color Colorless
Quantity 1 g
Solubility Information Fully miscible with water.
Formula Weight 359.18 (Anhydrous)
Concentration or Composition (by Analyte or Components) Pt 2% w/w (cont. Pt)
Physical Form Liquid
Chemical Name or Material Tetraammineplatinum (II) hydrogen phosphate solution
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'