Learn More
Tetraammineplatinum(II) nitrate solution, Pt 3-4% w/w (cont. Pt), Thermo Scientific Chemicals
172.86 € - 604.01 €
Chemical Identifiers
CAS | 20634-12-2 |
---|---|
Molecular Formula | H12N6O6Pt |
Molecular Weight (g/mol) | 387.22 |
MDL Number | MFCD00011624 |
InChI Key | RBAKORNXYLGSJB-UHFFFAOYSA-N |
Synonym | tetraammineplatinum ii nitrate, tetraammineplatinum dinitrate, tetrammineplatinum ii nitrate, tetraammineplatinum ii nitrate 1g, tetraammineplatinum ii nitrate, premion, platinum 2+ tetraamine dinitrate, tetraammineplatinum ii nitrate solution, pt w/w cont. pt |
PubChem CID | 146543 |
IUPAC Name | azane;platinum(2+);dinitrate |
SMILES | N.N.N.N.[Pt++].[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11345919
|
Thermo Scientific Alfa Aesar
012167.NJ |
0.5 g |
172.86 €
0.50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11335919
|
Thermo Scientific Alfa Aesar
012167.NA |
2 g |
604.01 €
2g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Tetraammineplatinum(II) nitrate is widely utilized as a precursor to bimetallic Pt/Zn catalysts for the selective hydrogenation of crotonaldehyde.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
20634-12-2 | |
387.22 | |
RBAKORNXYLGSJB-UHFFFAOYSA-N | |
146543 | |
N.N.N.N.[Pt++].[O-][N+]([O-])=O.[O-][N+]([O-])=O |
H12N6O6Pt | |
MFCD00011624 | |
tetraammineplatinum ii nitrate, tetraammineplatinum dinitrate, tetrammineplatinum ii nitrate, tetraammineplatinum ii nitrate 1g, tetraammineplatinum ii nitrate, premion, platinum 2+ tetraamine dinitrate, tetraammineplatinum ii nitrate solution, pt w/w cont. pt | |
azane;platinum(2+);dinitrate |
Specifications
20634-12-2 | |
Liquid | |
(NH3)4Pt(NO3)2 | |
UN3218 | |
Miscible with water. | |
N.N.N.N.[Pt++].[O-][N+]([O-])=O.[O-][N+]([O-])=O | |
387.22 | |
387.21 | |
1.05g/mL |
Colorless | |
H12N6O6Pt | |
MFCD00011624 | |
tetraammineplatinum ii nitrate, tetraammineplatinum dinitrate, tetrammineplatinum ii nitrate, tetraammineplatinum ii nitrate 1g, tetraammineplatinum ii nitrate, premion, platinum 2+ tetraamine dinitrate, tetraammineplatinum ii nitrate solution, pt w/w cont. pt | |
RBAKORNXYLGSJB-UHFFFAOYSA-N | |
azane;platinum(2+);dinitrate | |
146543 | |
Pt 3 to 4% w/w (cont. Pt) | |
Tetraammineplatinum(II) nitrate solution |
Safety and Handling
GHS H Statement
H240-H272
Heating may cause an explosion.
May intensify fire
oxidizer.
GHS P Statement
P221-P210-P370+P378a-P410-P411a+P235-P501a
Take any precaution to avoid mixing with combustibles.
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
In case of fire: Use for extinction: CO2, powder or water spray.
Protect from sunlight.
Store at temperatures not exceeding 392°F.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
missing translation for 'dotInformation' : Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: NITRATES, INORGANIC, AQUEOUS SOLUTION, N.O.S .
missing translation for 'einecsNumber' : 243-929-9
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.