missing translation for 'onlineSavingsMsg'
Learn More

Tetraammineplatinum(II) nitrate, Thermo Scientific Chemicals

176.88 € - 626.12 €

Chemical Identifiers

CAS 20634-12-2
Molecular Formula H12N6O6Pt
Molecular Weight (g/mol) 387.22
MDL Number MFCD00011624
InChI Key RBAKORNXYLGSJB-UHFFFAOYSA-N
Synonym tetraammineplatinum ii nitrate, tetraammineplatinum dinitrate, tetrammineplatinum ii nitrate, tetraammineplatinum ii nitrate 1g, tetraammineplatinum ii nitrate, premion, platinum 2+ tetraamine dinitrate, tetraammineplatinum ii nitrate solution, pt w/w cont. pt
PubChem CID 146543
IUPAC Name azane;platinum(2+);dinitrate
SMILES N.N.N.N.[Pt++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11329359
View Documents Promotion Details
Thermo Scientific Alfa Aesar
088960.03
1 g
176.88 €
1g
Please to purchase this item. Need a web account? Register with us today!
11319359
View Documents Promotion Details
Thermo Scientific Alfa Aesar
088960.06
5 g
626.12 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Tetraammineplatinum(II) nitrate is widely utilized as a precursor to bimetallic Pt/Zn catalysts for the selective hydrogenation of crotonaldehyde.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

20634-12-2
387.22
RBAKORNXYLGSJB-UHFFFAOYSA-N
146543
N.N.N.N.[Pt++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
H12N6O6Pt
MFCD00011624
tetraammineplatinum ii nitrate, tetraammineplatinum dinitrate, tetrammineplatinum ii nitrate, tetraammineplatinum ii nitrate 1g, tetraammineplatinum ii nitrate, premion, platinum 2+ tetraamine dinitrate, tetraammineplatinum ii nitrate solution, pt w/w cont. pt
azane;platinum(2+);dinitrate

Specifications

262°C (decomposition)
Chunks and/or Powder
MFCD00011624
Miscible with water.
N.N.N.N.[Pt++].[O-][N+]([O-])=O.[O-][N+]([O-])=O
387.22
387.21
20634-12-2
H12N6O6Pt
tetraammineplatinum ii nitrate, tetraammineplatinum dinitrate, tetrammineplatinum ii nitrate, tetraammineplatinum ii nitrate 1g, tetraammineplatinum ii nitrate, premion, platinum 2+ tetraamine dinitrate, tetraammineplatinum ii nitrate solution, pt w/w cont. pt
RBAKORNXYLGSJB-UHFFFAOYSA-N
azane;platinum(2+);dinitrate
146543
Tetraammineplatinum(II) nitrate
Videos
Safety and Handling

Safety and Handling

P210-P220-P221-P261-P264b-P271-P272-P280-P302+P352-P304+P340-P305+P351+P338-P312-P333+P313-P362-P370+P378q-P501c

H272-H315-H317-H319-H335

missing translation for 'dotInformation' : Hazard Class: 5.1; Packaging Group: II

missing translation for 'einecsNumber' : 243-929-9

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

Product Identifier
  • Tetraammineplatinum(II) nitrate

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Oxidising solid Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3

Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Supplemental information
  • MIXTURE LIST-Contains : Tetraammineplatinumdinitrate

SDS
Documents

Documents

Certificates

RUO – Research Use Only