missing translation for 'onlineSavingsMsg'
Learn More
Thermo Scientific Chemicals Tetrabromophenol Blue, pure Product Code.: 11388731

Thermo Scientific Chemicals Tetrabromophenol Blue, pure

Product Code. 11388731
25 g, Glass bottle
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 11388731

Brand: Thermo Scientific Chemicals 189910250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4430-25-5
Molecular Formula C19H6Br8O5S
Molecular Weight (g/mol) 985.53
InChI Key QPMIVFWZGPTDPN-UHFFFAOYSA-N
Synonym tetrabromophenol blue, 4,5,6,7-tetrabromo-3,3-bis 3,5-dibromo-4-hydroxyphenyl-3h-benzo c 1,2 oxathiole 1,1-dioxide, tetrabromphenol tetrabromsulfonphthalein, unii-s6fo6931n7, ccris 5488, 3',3,5',5-tetrabromophenol-4,5,6,7-tetrabromosulfonephthalein, phenol, 4,4'-4,5,6,7-tetrabromo-1,1-dioxido-3h-2,1-benzoxathiol-3-ylidene bis 2,6-dibromo, 4,4'-4,5,6,7-tetrabromo-3h-2,1-benzoxathiol-3-ylidene bis 2,6-dibromophenol s,s-dioxideShow MoreShow Less
PubChem CID 78159
IUPAC Name 2,6-dibromo-4-[4,5,6,7-tetrabromo-3-(3,5-dibromo-4-hydroxyphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]phenol
SMILES C1=C(C=C(C(=C1Br)O)Br)C2(C3=C(C(=C(C(=C3Br)Br)Br)Br)S(=O)(=O)O2)C4=CC(=C(C(=C4)Br)O)Br

Specifications

Melting Point 203.0°C to 205.0°C
Color Beige
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 25 g
Beilstein 19,II,107
Solubility Information Solubility in water: soluble in water.
Formula Weight 985.53
Grade Pure
Physical Form Powder
Chemical Name or Material Tetrabromophenol Blue
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0465087 Certificate TypeCertificate of Analysis Date12/08/2024 Catalog Number189910250,189910050
Lot NumberA0461354 Certificate TypeCertificate of Analysis Date25/03/2024 Catalog Number189910250,189910050
Lot NumberA0454361 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number189910250,189910050
Lot NumberA0450506 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number189910250,189910050
Lot NumberA0449065 Certificate TypeCertificate of Analysis Date09/01/2023 Catalog Number189910250,189910050
Product Identifier
  • Tetrabromophenol Blue

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Tetrabromophenol Blue, pure >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.