missing translation for 'onlineSavingsMsg'
Learn More

Tetrabromophthalic anhydride, 98%, Thermo Scientific Chemicals

Product Code. 11430388
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 632-79-1
Molecular Formula C8Br4O3
Molecular Weight (g/mol) 463.701
MDL Number MFCD00005919
InChI Key QHWKHLYUUZGSCW-UHFFFAOYSA-N
PubChem CID 12443
IUPAC Name 4,5,6,7-tetrabromo-2-benzofuran-1,3-dione
SMILES C12=C(C(=C(C(=C1Br)Br)Br)Br)C(=O)OC2=O

Specifications

Melting Point 274°C to 280°C
Density 2.87
Odor Odorless
Quantity 100 g
Beilstein 18908
Sensitivity Moisture sensitive
Synonym tetrabromophthalic anhydride, bromphthal, firemaster pht 4, phthalic anhydride, tetrabromo, bromophthal, tetrabromophthalic acid anhydride, firemaster pht4, 1,3-isobenzofurandione, 4,5,6,7-tetrabromo, 4,5,6,7-tetrabromo-1,3-isobenzofurandione, pht 4
Formula Weight 463.7
Percent Purity 98%
Chemical Name or Material Tetrabromophthalic anhydride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'