Learn More
Tetrabutylammonium bromide, 99+%, Thermo Scientific Chemicals
28.04 € - 838.17 €
Chemical Identifiers
CAS | 1643-19-2 |
---|---|
Molecular Formula | C16H36BrN |
Molecular Weight (g/mol) | 322.36 |
MDL Number | MFCD00011633 |
InChI Key | JRMUNVKIHCOMHV-UHFFFAOYSA-M |
Synonym | tetrabutylammonium bromide, tetra-n-butylammonium bromide, tbab, tetrabutylazanium bromide, tetrabutyl ammonium bromide, tetrabutyl-ammonium bromide, aliquat 100, tbabr, ipc-tba-br, unii-vjz168i98rShow More |
PubChem CID | 74236 |
ChEBI | CHEBI:51993 |
IUPAC Name | tetrabutylazanium;bromide |
SMILES | CCCC[N+](CCCC)(CCCC)CCCC.[Br-] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10646202
|
Thermo Scientific Acros
185680250 |
25 g | Glass bottle |
28.04 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10586783
|
Thermo Scientific Acros
185680500 |
50 g | Glass bottle |
42.41 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10327940
|
Thermo Scientific Acros
185681000 |
100 g | Glass bottle |
72.66 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10266000
|
Thermo Scientific Acros
185685000 |
500 g | Glass bottle |
243.54 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11345932
|
Thermo Scientific Acros
185680025 |
2.5 kg | Plastic bottle |
838.17 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalystChemical Identifiers
1643-19-2 | |
322.36 | |
JRMUNVKIHCOMHV-UHFFFAOYSA-M | |
74236 | |
tetrabutylazanium;bromide |
C16H36BrN | |
MFCD00011633 | |
tetrabutylammonium bromide, tetra-n-butylammonium bromide, tbab, tetrabutylazanium bromide, tetrabutyl ammonium bromide, tetrabutyl-ammonium bromide, aliquat 100, tbabr, ipc-tba-br, unii-vjz168i98rShow More | |
CHEBI:51993 | |
CCCC[N+](CCCC)(CCCC)CCCC.[Br-] |
Specifications
1643-19-2 | |
Cream to White | |
99% min. (Argentometry) | |
[CH3(CH2)3]4NBr | |
04,477; 05,644; 07,353; 17,363 | |
Solubility in water: 600g/L (20°C). | |
CCCC[N+](CCCC)(CCCC)CCCC.[Br-] | |
322.36 | |
CHEBI:51993 | |
99+% | |
Tetrabutylammonium bromide, 99+% |
100.0°C to 104.0°C | |
Authentic | |
C16H36BrN | |
MFCD00011633 | |
tetrabutylammonium bromide, tetra-n-butylammonium bromide, tbab, tetrabutylazanium bromide, tetrabutyl ammonium bromide, tetrabutyl-ammonium bromide, aliquat 100, tbabr, ipc-tba-br, unii-vjz168i98r | |
JRMUNVKIHCOMHV-UHFFFAOYSA-M | |
tetrabutylazanium;bromide | |
74236 | |
322.36 | |
Crystalline Powder |
Safety and Handling
GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 216-699-2
missing translation for 'tsca' : TSCA
- Tetrabutylammonium bromide
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
- MIXTURE LIST-Contains : Tetrabutylammonium bromide
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.