missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylammonium hydroxide, 0.1N sol. in toluene/methanol for non-aq.titr., Thermo Scientific Chemicals

Tetrabutylammonium hydroxide, 0.1N sol. in toluene/methanol for non-aq.titr., Acros Organics, Quantity: 500mL, Packaging: Glass bottle, Colorless to Yellow, Molecular Weight: 259.47, Beilstein: 04,II,634, CAS: 2052-49-5, 108-88-3, 67-56-1, Density: 0.8500g/mL, EINECS Number: 218-147-6

51.66 € - 505.52 €

Specifications

Chemical Name or Material Tetrabutylammonium hydroxide
CAS 2052-49-5, 108-88-3, 67-56-1
Color Colorless to Yellow
Density 0.8500g/mL
Flash Point 4°C
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10564721
View Documents Promotion Details
Thermo Scientific Acros
123531000
100 mL Glass bottle
51.66 €
100mL
Please to purchase this item. Need a web account? Register with us today!
10551751
View Documents Promotion Details
Thermo Scientific Acros
123535000
500 mL Glass bottle
153.77 €
500mL
Please to purchase this item. Need a web account? Register with us today!
11955441
View Documents Promotion Details
Thermo Scientific Acros
123530025
2.5 L Glass bottle
505.52 €
2.50 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
Specifications

Specifications

Tetrabutylammonium hydroxide
Colorless to Yellow
4°C
C16H37NO
1.3775
04,II,634
0.85
Solubility in water: insoluble
[OH-].CCCC[N+](CCCC)(CCCC)CCCC
259.48
259.47
2052-49-5, 108-88-3, 67-56-1
0.8500g/mL
Glass bottle
[CH3(CH2)3]4NOH
MFCD00009425
05,645; 11,500; 07,332
tetrabutylammonium hydroxide, tetra-n-butylammonium hydroxide, tetrabutylazanium hydroxide, tetrabutylammoniumhydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide, ammonium, tetrabutyl-, hydroxide, n,n,n-tributyl-1-butanaminium hydroxide, tetra n-butyl ammonium hydroxide, tetra-n-butyl ammonium hydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide 1:1
VDZOOKBUILJEDG-UHFFFAOYSA-M
tetrabutylazanium;hydroxide
2723671
Solution
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Highly flammable liquid and vapor.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes damage to organs.
Suspected of damaging the unborn child.
May be fatal if swallow

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF SWALLOWED: rinse mouth.
Do NOT induce vomi

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 218-147-6

Product Identifier
  • Tetrabutylammonium hydroxide, 0.1N sol. in toluene/methanol

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Aspiration toxicity Category 1
  • Specific target organ toxicity Category 3
  • Flammable liquid Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity after single exposure Category 1
  • Serious eye damage/eye irritation Category 1
  • Acute toxicity Category 4

Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H361d-Suspected of damaging the unborn child.
  • H373-May cause damage to organs.
  • H332-Harmful if inhaled.
  • H312-Harmful in contact with skin.
  • H370-Causes damage to organs.
  • H225-Highly flammable liquid and vapour.
  • H336-May cause drowsiness or dizziness.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.,Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.,IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P331-Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only