missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylammonium hydroxide 30-hydrate, 95+%, Thermo Scientific Chemicals

Product Code. 11915461
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 147741-30-8
Molecular Formula C16H97NO31
Molecular Weight (g/mol) 799.93
MDL Number MFCD00149573
InChI Key DFGIRVKFXSRUOX-UHFFFAOYSA-M
Synonym tetrabutylammonium hydroxide 30-hydrate, tetrabutylammonium ion triacontahydrate hydroxide, n,n,n-tributylbutan-1-aminium hydroxide-water 1/1/30
PubChem CID 16218633
SMILES O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.[OH-].CCCC[N+](CCCC)(CCCC)CCCC

Specifications

Melting Point 27.0°C to 30.0°C
Color White
Physical Form Crystals
Quantity 100 g
Assay Percent Range 95+%
IUPAC Name tetrabutylazanium triacontahydrate hydroxide
Formula Weight 799.93
Percent Purity ≥95%
Packaging Glass bottle
Chemical Name or Material Tetrabutylammonium hydroxide 30-hydrate
compliance-icons
Product Identifier
  • Tetrabutylammonium hydroxide 30-hydrate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Tetrabutylammonium hydroxide 30-hydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.