missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrabutylammonium Hydroxide Solution, Volumetric, 0.1M in Toluene/Methanol (0.1N), Honeywell Fluka™
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle
Description
Honeywell´s high-quality titration products cover the complete titration workflow and include:
- Solutions for volumetric titration (acids, bases, or salts)
- Buffers available as concentrates or ready-to-use
- Reagents for complexometry:
- Aminopolycarboxylic acids (EDTA/NTA analogs)
- Masking agents
- Indicators for volumetric and complexometric titrations
Specifications
Specifications
Color | Colorless |
CAS | 2052-49-5 |
Boiling Point | 64°C |
Molecular Formula | C16H37NO |
Linear Formula | (CH3CH2CH2CH2)4N(OH) |
MDL Number | MFCD00009425 |
UN Number | UN1992 |
Beilstein | 3571228 |
Synonym | tetrabutylammonium hydroxide, tetra-n-butylammonium hydroxide, tetrabutylazanium hydroxide, tetrabutylammoniumhydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide, ammonium, tetrabutyl-, hydroxide, n,n,n-tributyl-1-butanaminium hydroxide, tetra n-butyl ammonium hydroxide, tetra-n-butyl ammonium hydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide 1:1 |
Solubility Information | Soluble in water |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'