missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylammonium Hydroxide Titrant, 0.4M in Water, HPLC Grade, Thermo Scientific Chemicals

Product Code. 10055680
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ion pair chromatography, Phase transfer catalyst
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2052-49-5
Molecular Formula C16H37NO
Molecular Weight (g/mol) 259.48
MDL Number MFCD00009425
InChI Key VDZOOKBUILJEDG-UHFFFAOYSA-M
Synonym tetrabutylammonium hydroxide, tetra-n-butylammonium hydroxide, tetrabutylazanium hydroxide, tetrabutylammoniumhydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide, ammonium, tetrabutyl-, hydroxide, n,n,n-tributyl-1-butanaminium hydroxide, tetra n-butyl ammonium hydroxide, tetra-n-butyl ammonium hydroxide, 1-butanaminium, n,n,n-tributyl-, hydroxide 1:1
PubChem CID 2723671
SMILES [OH-].CCCC[N+](CCCC)(CCCC)CCCC

Specifications

Absorbance (1 cm cell vs water), (1 cm cell vs water) at 0nm, 0.15 max. at 420nm
Color Undesignated
Linear Formula (CH3(CH2)3)4NOH
Beilstein 04, II, 634
Concentration 0.37 to 0.43M
Fieser 05,645; 11,500
Density 0.85
Solubility Information Solubility in water: soluble.
IUPAC Name tetrabutylazanium hydroxide
Formula Weight 259.47
Grade HPLC
Packaging Plastic bottle
Quantity 100 g
Flash Point >110°C
Physical Form Solution
Chemical Name or Material Tetrabutylammonium hydroxide titrant, 0.4M in Water
Show More Show Less
compliance-icons
Product Identifier
  • Tetrabutylammonium hydroxide, 0.4M in water
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'