missing translation for 'onlineSavingsMsg'
Learn More
Tetrabutylammonium nitrate, 98%, Thermo Scientific Chemicals Product Code.: 10617221

Tetrabutylammonium nitrate, 98%, Thermo Scientific Chemicals

Product Code. 10617221
25 g, Glass bottle
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10617221

Brand: Thermo Scientific Acros 420130250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1941-27-1
Molecular Formula C16H36N2O3
Molecular Weight (g/mol) 304.46
MDL Number MFCD00043202
InChI Key QHOKENWFMZXSEU-UHFFFAOYSA-N
Synonym tetrabutylammonium nitrate, tetra-n-butylammonium nitrate, 1-butanaminium, n,n,n-tributyl-, nitrate, tetrabutylazanium nitrate, ammonium, tetrabutyl-, nitrate, n-n-n-tributyl-1-butanaminium nitrate, 1-butanaminium, n,n,n-tributyl-, nitrate 1:1, tetrabutylammonium ion nitrate, tetrabutylammoniumnitrate, acmc-209exx
PubChem CID 16027
IUPAC Name tetrabutylazanium;nitrate
SMILES CCCC[N+](CCCC)(CCCC)CCCC.[N+](=O)([O-])[O-]

Specifications

Melting Point 116°C to 120°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula (CH3(CH2)3)4NNO3
Quantity 25 g
Solubility Information Solubility in water: <2%
Formula Weight 304.46
Percent Purity 98%
Physical Form Fine Crystalline Powder
Chemical Name or Material Tetrabutylammonium nitrate, 98%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0466656 Certificate TypeCertificate of Analysis Date09/10/2024 Catalog Number420130050,420131000,420130250
Lot NumberA0463685 Certificate TypeCertificate of Analysis Date18/06/2024 Catalog Number420130050,420131000,420130250
Lot NumberA0463684 Certificate TypeCertificate of Analysis Date14/06/2024 Catalog Number420130050,420131000,420130250
Lot NumberA0460152 Certificate TypeCertificate of Analysis Date14/02/2024 Catalog Number420130050,420131000,420130250
Lot NumberA0460153 Certificate TypeCertificate of Analysis Date13/02/2024 Catalog Number420130050,420131000,420130250
compliance-icons compliance-icons
Product Identifier
  • Tetrabutylammonium nitrate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Oxidising solid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H272-May intensify fire; oxidiser.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tetrabutylammonium nitrate, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.