missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylammonium perchlorate, 98%

Product Code. 10568902 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 10568902

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1923-70-2 | C16H36ClNO4 | 341.917 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent, Phase transfer catalyst
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1923-70-2
Molecular Formula C16H36ClNO4
Molecular Weight (g/mol) 341.917
MDL Number MFCD00038722
InChI Key KBLZDCFTQSIIOH-UHFFFAOYSA-M
Synonym tetrabutylammonium perchlorate, 1-butanaminium, n,n,n-tributyl-, perchlorate, tetrabutyl ammonium perchlorate, tetra-n-butylammonium perchlorate, tetrabutylammonium perchlorat, 1-butanaminium, n,n,n-tributyl-, perchlorate 1:1, acmc-1bpvk, n,n,n-tributylbutan-1-aminium perchlorate, tetrabutylammoniumperchlorate, tetrabutylazanium perchlorate
PubChem CID 74723
IUPAC Name tetrabutylazanium;perchlorate
SMILES CCCC[N+](CCCC)(CCCC)CCCC.[O-]Cl(=O)(=O)=O

Specifications

Color White
Linear Formula (CH3(CH2)3)4NClO4
Formula Weight 341.91
Percent Purity ≥97.5%
Packaging Glass bottle
Decomposition Information 290°C
Quantity 100 g
Physical Form Crystalline Powder
Chemical Name or Material Tetrabutylammonium perchlorate, Contains Max. 10% Water
compliance-icons compliance-icons
Product Identifier
  • Tetrabutylammonium perchlorate
Signal Word
  • Danger
Hazard Category
  • Oxidising solid Category 2
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P371+P380+P375-In case of major fire and large quantities: Evacuate area Fight fire remotely due to the risk of explosion.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.