Learn More
Tetrabutylammonium phosphate monobasic, 0.5M conc. in HPLC grade water, HPLC grade, Thermo Scientific Chemicals
51.36 € - 413.06 €
Specifications
Boiling Point | 100.0°C |
---|---|
Chemical Name or Material | Tetrabutylammonium phosphate monobasic |
CAS | 5574-97-0, 7732-18-5 |
Physical Form | Solution |
Molecular Formula | C16H38NO4P |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10526412
|
Thermo Scientific Acros
420170250 |
25 mL | Glass bottle |
51.36 €
25mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10512204
|
Thermo Scientific Acros
420171000 |
100 mL | Glass bottle |
116.58 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11935451
|
Thermo Scientific Acros
420175000 |
500 mL | Glass bottle |
413.06 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ion pair chromatography, Phase transfer catalystSpecifications
100.0°C | |
5574-97-0, 7732-18-5 | |
C16H38NO4P | |
MFCD00064526 | |
ARRNBPCNZJXHRJ-UHFFFAOYSA-M | |
dihydrogen phosphate;tetrabutylazanium | |
2735142 | |
0.48 to 0.52M | |
1.01 |
Tetrabutylammonium phosphate monobasic | |
Solution | |
(CH3(CH2)3)4NH2PO4 | |
tetrabutylammonium dihydrogen phosphate, tetrabutylammonium phosphate, tetrabutylammonium dihydrogenphosphate, 1-butanaminium, n,n,n-tributyl-, phosphate 1:1, tetra-n-butylammonium phosphate, tetrabutylazanium dihydrogen phosphate, dihydrogen phosphate; tetrabutylazanium, dihydrogen phosphate; tetrabutylammonium, tetrabutyl ammonium dihydrogen phosphate, tetrabutylammonium ion dihydrogenphosphate ion | |
OP(O)([O-])=O.CCCC[N+](CCCC)(CCCC)CCCC | |
339.46 | |
339.45 | |
Glass bottle | |
1.0100g/mL |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
missing translation for 'einecsNumber' : 226-947-1
- Tetrabutylammonium phosphate, 0.5M concentrate in HPLC grade water
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.