missing translation for 'onlineSavingsMsg'
Learn More
Tetrabutylammonium phosphate monobasic, 99%, HPLC grade, Thermo Scientific Chemicals Product Code.: 10569092

Tetrabutylammonium phosphate monobasic, 99%, HPLC grade, Thermo Scientific Chemicals

Product Code. 10569092
5 g, Glass bottle
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10569092

Brand: Thermo Scientific Acros 420160050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ion pair chromatography, Phase transfer catalyst
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5574-97-0
Molecular Formula C16H38NO4P
Molecular Weight (g/mol) 339.46
MDL Number MFCD00064526
InChI Key ARRNBPCNZJXHRJ-UHFFFAOYSA-M
Synonym tetrabutylammonium dihydrogen phosphate, tetrabutylammonium phosphate, tetrabutylammonium dihydrogenphosphate, 1-butanaminium, n,n,n-tributyl-, phosphate 1:1, tetra-n-butylammonium phosphate, tetrabutylazanium dihydrogen phosphate, dihydrogen phosphate; tetrabutylazanium, dihydrogen phosphate; tetrabutylammonium, tetrabutyl ammonium dihydrogen phosphate, tetrabutylammonium ion dihydrogenphosphate ion
PubChem CID 2735142
IUPAC Name dihydrogen phosphate;tetrabutylazanium
SMILES OP(O)([O-])=O.CCCC[N+](CCCC)(CCCC)CCCC

Specifications

Absorbance (0.1% aq. soln.) (1 cm quartz cell), (0.1% aq. soln.) (1 cm quartz cell) at 0nm, 0.05 max. at 214nm, 0.1 max. at 210nm, 0.2 max. at 200nm
Melting Point 148.0°C to 151.0°C
Color Gray to Black
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula (CH3(CH2)3)4NH2PO4
Quantity 5 g
Solubility Information Solubility in water: soluble.
Formula Weight 339.45
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Tetrabutylammonium phosphate monobasic, 99%
Show More Show Less
Product Identifier
  • Tetrabutylammonium phosphate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tetrabutylammonium phosphate monobasic, 99%, HPLC grade, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.