missing translation for 'onlineSavingsMsg'
Learn More

Tetrabutylphosphonium hydroxide, 40 wt.% solution in water, Thermo Scientific Chemicals

 

Brand:  Thermo Scientific Acros 395031000

105.88 EUR valid until 2024-12-31
Use promo code "21665" to get your promotional price.



Product Code. 11955871

  • 119.60 € / 100mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For C-C Bond Formation (Olefination)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Tetrabutylphosphonium hydroxide
15.0°C
0.9990g/mL
C16H37OP
100 mL
tetrabutylphosphonium hydroxide, tbph, tetrabutylphosphonium hydroxide solution, phosphonium,tetrabutyl-, hydroxide 1:1, phosphonium, tetrabutyl-, hydroxide 1:1, acmc-1c8ln, tetrabutylphosphoniumhydroxide, tetrabutylphosphanium hydroxide, tetra-n-butylphosphonium hydroxide w/w aq. soln., tetrabutylphosphonium hydroxide solution, 40 wt. % in h2o
DFQPZDGUFQJANM-UHFFFAOYSA-M
tetrabutylphosphanium;hydroxide
84487
37.5 to 42.5%
14518-69-5, 7732-18-5
Colorless to Yellow
Plastic Bottle
MFCD00068456
0.999
Solubility in water: soluble.
[OH-].CCCC[P+](CCCC)(CCCC)CCCC
276.45
276.44
Liquid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Tetrabutylphosphonium hydroxide, 40 wt.% solution in water

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

RUO – Research Use Only