missing translation for 'onlineSavingsMsg'
Learn More

Tetracaine, 98%, Thermo Scientific Chemicals

43.00 € - 391.95 €

Chemical Identifiers

CAS 94-24-6
Molecular Formula C15H24N2O2
Molecular Weight (g/mol) 264.36
InChI Key GKCBAIGFKIBETG-UHFFFAOYSA-N
PubChem CID 5411
ChEBI CHEBI:9468
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15751759
View Documents Promotion Details
Thermo Scientific Acros
460820050
5 g
43.00 €
5g
Please to purchase this item. Need a web account? Register with us today!
15761759
View Documents Promotion Details
Thermo Scientific Acros
460820250
25 g
107.54 €
25g
Please to purchase this item. Need a web account? Register with us today!
15771759
View Documents Promotion Details
Thermo Scientific Acros
460821000
100 g
391.95 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

94-24-6
264.36
5411
C15H24N2O2
GKCBAIGFKIBETG-UHFFFAOYSA-N
CHEBI:9468

Specifications

94-24-6
C15H24N2O2
GKCBAIGFKIBETG-UHFFFAOYSA-N
5411
264.36
Tetracaine
43°C
Solubility in water: .. Other solubilities: soluble in methanol
264.36
CHEBI:9468
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 202-316-6

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only