missing translation for 'onlineSavingsMsg'
Learn More
Tetrachlorobis(tetrahydrofuran)titanium(IV), 98%, Thermo Scientific Chemicals Product Code.: 11304517

Tetrachlorobis(tetrahydrofuran)titanium(IV), 98%, Thermo Scientific Chemicals

Product Code. 11304517
10 g
Click to view available options
Quantity:
2 g
10 g
50 g
This item is not returnable. View return policy

Product Code. 11304517

Brand: Thermo Scientific Alfa Aesar 039594.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31011-57-1
Molecular Formula C8H16Cl4O2Ti
Molecular Weight (g/mol) 333.881
MDL Number MFCD00077884
InChI Key LXWBMENBONGPSB-UHFFFAOYSA-J
Synonym tetrachlorobis tetrahydrofuran titanium, tetrachlorobis tetrahydrofuran titanium iv, bis tetrahydrofuran ; titanium tetrachloride, titanium iv chloride tetrahydrofuran complex
PubChem CID 10958563
IUPAC Name oxolane;tetrachlorotitanium
SMILES C1CCOC1.C1CCOC1.Cl[Ti](Cl)(Cl)Cl

Specifications

Melting Point 126°C to 128°C
Color Yellow
Flash Point -17°C (1°F)
Quantity 10 g
UN Number UN2925
Sensitivity Moisture sensitive
Solubility Information Reacts with water
Formula Weight 333.91
Percent Purity 98%
Physical Form Crystalline or Powder
Chemical Name or Material Tetrachlorobis(tetrahydrofuran)titanium(IV)
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP04F037 Certificate TypeCertificate of Analysis Date13/01/2025 Catalog Number039594.09
Lot NumberR17I037 Certificate TypeCertificate of Analysis Date02/08/2022 Catalog Number039594.04,039594.09
Lot NumberW21E049 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number039594.04,039594.09
Lot NumberB06P31 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number039594.04,039594.09
Lot NumberP04F037 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number039594.04,039594.09

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tetrachlorobis(tetrahydrofuran)titanium(IV), 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.