missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrachloroethylene, CHROMASOLV™, for HPLC, ≥99.9%, Honeywell Riedel-de Haen
Click to view available options
Quantity:
100 mL
1 L
Packaging:
Glass Bottle
Description
- For HPLC and organic synthesis
- High-purity, multipurpose solvents
- High lot-to-lot consistency
Specifications
Specifications
Absorbance | max. 1.0 at 290 nm, max. 0.03 at 400 nm, max. 0.05 at 350 nm, max. 0.10 at 305 nm, max. 0.20 at 300 nm, max. 0.30 at 295 nm |
CAS Max % | 100.0000% |
Melting Point | -22°C (literature) |
Density | 1.623 g/mL at 25 °C(lit.) |
Boiling Point | 120°C to 122°C |
Packaging | Glass Bottle |
Linear Formula | CCl2=CCl2 |
Quantity | 1 L |
UN Number | UN1897 |
Beilstein | 1361721 |
Show More |
Safety and Handling
Product Identifier
- Tetrachloroethylene
- Warning
- LONG-TERM AQUATIC HAZARD Chronic 2
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- H351-Suspected of causing cancer.
- H411-Toxic to aquatic life with long lasting effects.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : tetrachloroethylene
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction