missing translation for 'onlineSavingsMsg'
Learn More

Tetrachloroethylene, Honeywell

Product Code. 15661550
Click to view available options
Quantity:
1 L
2.5 L
10 L
800 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 127-18-4
Molecular Formula C2Cl4
Molecular Weight (g/mol) 165.82
MDL Number MFCD00000834
InChI Key CYTYCFOTNPOANT-UHFFFAOYSA-N
Synonym tetrachloroethylene, tetrachloroethene, perchloroethylene, perchlorethylene, perc, ethene, tetrachloro, tetrachlorethylene, ethylene tetrachloride, carbon dichloride, ankilostin
PubChem CID 31373
ChEBI CHEBI:17300
SMILES ClC(Cl)=C(Cl)Cl

Specifications

Density 1.623g/mL at 25°C
Linear Formula CCl2=CCl2
Quantity 800 kg
UN Number 1897
Beilstein 1361721
IUPAC Name tetrachloroethene
Vapor Density 5.83 (vs air)
Formula Weight 165.83g/mol
Vapor Pressure 13mmHg (20°C), 19mmHg (25°C)
Percent Purity ≥99.5%
Chemical Name or Material Tetrachloroethylene
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.