missing translation for 'onlineSavingsMsg'
Learn More
Tetrachlorophthalic anhydride, 98%, Thermo Scientific Chemicals Product Code.: 11477997

Tetrachlorophthalic anhydride, 98%, Thermo Scientific Chemicals

Product Code. 11477997
500 g
missing translation for 'orderingAttributeHoverText'
Quantity:
10,000 g
2500 g
500 g
This item is not returnable. View return policy

Product Code. 11477997

Brand: Thermo Scientific Alfa Aesar A17404.36

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Protection of amines as their tetrachlorophthalimide derivatives, by reaction with the anhydride in the presence of triethylamine, has been found to be useful in oligosaccharide chemistry. The group is stable to a wide range of reagents but can be selectively cleaved with ethylenediamine at lower temperatures than for the parent phthalimide derivatives; in particular, carboxylic esters are not cleaved under these conditions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Protection of amines as their tetrachlorophthalimide derivatives, by reaction with the anhydride in the presence of triethylamine, has been found to be useful in oligosaccharide chemistry. The group is stable to a wide range of reagents but can be selectively cleaved with ethylenediamine at lower temperatures than for the parent phthalimide derivatives; in particular, carboxylic esters are not cleaved under these conditions.

Solubility
Hydrolyzes slowly in water.

Notes
Hygroscopic. Store away from oxidizing agents, acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 117-08-8
Molecular Formula C8Cl4O3
Molecular Weight (g/mol) 285.885
MDL Number MFCD00005920
InChI Key AUHHYELHRWCWEZ-UHFFFAOYSA-N
Synonym tetrachlorophthalic anhydride, tetrathal, 4,5,6,7-tetrachloro-1,3-isobenzofurandione, niagathal, 1,3-isobenzofurandione, 4,5,6,7-tetrachloro, phthalic anhydride, tetrachloro, 4,5,6,7-tetrachloroisobenzofuran-1,3-dione, 1,3-dioxy-4,5,6,7-tetrachloroisobenzofuran, unii-76glw0lbek, ccris 6202
PubChem CID 8326
ChEBI CHEBI:59097
IUPAC Name 4,5,6,7-tetrachloro-2-benzofuran-1,3-dione
SMILES C12=C(C(=C(C(=C1Cl)Cl)Cl)Cl)C(=O)OC2=O

Specifications

Melting Point 254°C to 258°C
Boiling Point 349°C to 354°C
Flash Point 362°C (683°F)
Odor Weak
Quantity 500 g
UN Number UN3077
Beilstein 211560
Sensitivity Moisture sensitive
Solubility Information Hydrolyzes slowly in water.
Formula Weight 285.9
Percent Purity 98%
Chemical Name or Material Tetrachlorophthalic anhydride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10214822 Certificate TypeCertificate of Analysis Date20/07/2023 Catalog NumberA17404,A17404.0C,A17404.36,A17404.0E
Lot Number10210338 Certificate TypeCertificate of Analysis Date20/07/2023 Catalog NumberA17404,A17404.0C,A17404.36,A17404.0E
Lot Number10166645 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA17404.0C,A17404.36,A17404.0E
Lot Number10158320 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA17404.0C,A17404.36,A17404.0E
Lot Number10214822 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA17404.0C,A17404.36,A17404.0E

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tetrachlorophthalic anhydride, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.