Learn More
Gibco™ Tetracycline Hydrochloride
Tetracycline is a yellow, odorless, crystalline powder used as an agent to activate gene expression from an inducible expression plasmid in engineered cells expressing the tetracycline repressor protein
Brand:  Gibco™ A39246
Product Code. 16376552
  • 65.25 € / Each
Explore more special offers
Description
Description
Tetracycline Hydrochloride is the hydrochloride salt of tetracycline
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Tetracycline Hydrochloride
Powder
500 mg
Wet Ice