missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Tetracycline hydrochloride, can be used as secondary standard

Product Code. 10695272
Click to view available options
Quantity:
500 mg
2.5 g
Packaging:
Glass Bottle

Tetracycline hydrochloride, CAS # 64-75-5, is a wide-spectrum antibiotic that targets both gram-negative and gram-positive bacteria.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description
• Tetracycline hydrochloride is the hydrochloride salt of
the antibiotic tetracycline
• Tetracycline blocks amino acyl-tRNA binding during
bacterial protein synthesis
• Tetracycline is a broad-spectrum naphthacene antibiotic that
inhibits growth of both gram positive and gram negative bacteria
Applications
• Tetracycline may be used as a selection agent in cell
culture
• It may also be used to activate inducible gene
expression in engineered cells
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-75-5
Molecular Formula C22H25ClN2O8
Molecular Weight (g/mol) 480.898
MDL Number MFCD00078142
InChI Key HTXDZWDXSWLLLW-FMZCEJRJSA-N
PubChem CID 129628373
IUPAC Name (4S,4aS,5aS,6S,12aS)-4-(dimethylamino)-3,6,10,11,12a-pentahydroxy-6-methyl-1,12-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide;hydrochloride
SMILES CC1(C2CC3C(C(=C(C(=O)C3(C(=O)C2=C(C4=C1C=CC=C4O)O)O)C(=O)N)O)N(C)C)O.Cl

Specifications

Type Tetracycline Hydrochloride
Physical Form Powder
Melting Point 223.0°C
Infrared Spectrum Authentic
Packaging Glass Bottle
Specific Rotation − 252.00
Color Yellow
Quantity 500 mg
Beilstein 14, IV, 2625
Merck Index 15, 9341
Solubility Information Solubility in water: 20g/L. Other solubilities: soluble in ethanol: 5g/L, insoluble in ether, hydrocarbons and acetone
Formula Weight 480.9
Percent Purity ≥95 (anhydrous and solvent free), 80 wt% (as is, calculated)
Chemical Name or Material Tetracycline hydrochloride
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Tetracycline hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 2
Hazard Statement
  • H361d-Suspected of damaging the unborn child.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.