Learn More
Tetradecanonitrile, 98%, Thermo Scientific™
24.62 € - 365.00 €
Chemical Identifiers
CAS | 629-63-0 |
---|---|
Molecular Formula | C14H27N |
Molecular Weight (g/mol) | 209.38 |
MDL Number | MFCD00019848 |
InChI Key | MLRCLPRHEOPXLL-UHFFFAOYSA-N |
Synonym | myristonitrile, tetradecanonitrile, 1-cyanotridecane, myristyl nitrile, n-tridecyl cyanide, myrystyl nitrile, tridecyl cyanide, c14-18 nitriles, c14-c18 alkylnitrile, acmc-209nb3 |
PubChem CID | 64678 |
IUPAC Name | tetradecanenitrile |
SMILES | CCCCCCCCCCCCCC#N |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
629-63-0 | |
209.38 | |
MLRCLPRHEOPXLL-UHFFFAOYSA-N | |
64678 | |
CCCCCCCCCCCCCC#N |
C14H27N | |
MFCD00019848 | |
myristonitrile, tetradecanonitrile, 1-cyanotridecane, myristyl nitrile, n-tridecyl cyanide, myrystyl nitrile, tridecyl cyanide, c14-18 nitriles, c14-c18 alkylnitrile, acmc-209nb3 | |
tetradecanenitrile |
Specifications
629-63-0 | |
0.83 | |
Odorless | |
1.4415 | |
1705598 | |
MLRCLPRHEOPXLL-UHFFFAOYSA-N | |
tetradecanenitrile | |
64678 | |
98% |
∼19°C | |
226°C to 227°C (100 mmHg) | |
C14H27N | |
MFCD00019848 | |
myristonitrile, tetradecanonitrile, 1-cyanotridecane, myristyl nitrile, n-tridecyl cyanide, myrystyl nitrile, tridecyl cyanide, c14-18 nitriles, c14-c18 alkylnitrile, acmc-209nb3 | |
CCCCCCCCCCCCCC#N | |
209.38 | |
209.38 | |
Tetradecanonitrile |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
P264b-P270-P301+P312-P330-P501c
H302
missing translation for 'einecsNumber' : 211-099-7
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
Your input is important to us. Please complete this form to provide feedback related to the content on this product.