missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetraethylammonium bromide, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Plastic Drum
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalyst
Specifications
Specifications
Melting Point | 285.0°C |
Assay Percent Range | 97.5% min. (Argentometry) |
Packaging | Plastic bottle |
Linear Formula | (C2H5)4NBr |
Quantity | 1 kg |
Beilstein | 04,104 |
Fieser | 06,568; 13,46 |
Merck Index | 15,9344 |
Solubility Information | Solubility in water: 2795g/L (25°C). Other solubilities: freely soluble in alcohol,chloroform and acetone,slightly soluble in benzene |
Formula Weight | 210.16 |
Show More |
Safety and Handling
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 3
- H412-Harmful to aquatic life with long lasting effects.
- P273-Avoid release to the environment.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction