missing translation for 'onlineSavingsMsg'
Learn More

Tetraethylammonium chloride, Tocris Bioscience™

Product Code. 18094412
Click to view available options
Quantity:
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Non-selective K+ channel blocker

Non-selective K+ channel blocker.

Specifications

CAS 56-34-8
Molecular Formula C8H20ClN
MDL Number MFCD00149992
Quantity 50 mg
Synonym tetraethylammonium chloride, tetraethyl ammonium chloride, etamon chloride, tea chloride, teac, n,n,n-triethylethanaminium chloride, tetraethylammoniumchloride, ethanaminium, n,n,n-triethyl-, chloride, tetraethyl-ammonium chloride, unii-8b82z83xnn
InChI Key YMBCJWGVCUEGHA-UHFFFAOYSA-M
SMILES [Cl-].CC[N+](CC)(CC)CC
IUPAC Name tetraethylazanium chloride
Molecular Weight (g/mol) 165.71
PubChem CID 5946
ChEBI CHEBI:78161
Formula Weight 165.2
Percent Purity >98%
Chemical Name or Material Tetraethylammonium chloride
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.