missing translation for 'onlineSavingsMsg'
Learn More

Tetraethylammonium Hydroxide, 10% in Water, Thermo Scientific Chemicals

Brand:  Thermo Scientific Acros 420281000

29.17 EUR valid until 2024-10-31
Use promo code "21692" to get your promotional price.


 View more versions of this product

Product Code. 10016000

  • 30.70 € / 100g

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Tetraethylammonium hydroxide
10% in Water
102.0°C
(C2H5)4NOH
1.0200g/mL
tetraethylammonium hydroxide, tetraethyl ammonium hydroxide, unii-ra8vu41b1f, ammonium, tetraethyl-, hydroxide, tetraethylazanium hydroxide, ethanaminium, n,n,n-triethyl-, hydroxide, ra8vu41b1f, ethanaminium, n,n,n-triethyl-, hydroxide 1:1, tetraethylammonium hydroxide solution in water, n,n,n-triethylethanaminium
LRGJRHZIDJQFCL-UHFFFAOYSA-M
tetraethylazanium;hydroxide
6509
9.5 to 10.5%
1.402
100 g
Colorless
77-98-5, 7732-18-5
C8H21NO
MFCD00009024
15, 9346
Solubility in water: soluble.
[OH-].CC[N+](CC)(CC)CC
147.26
147.26
Plastic bottle
1.02
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

RUO – Research Use Only