Learn More
Tetraethylammonium hydroxide, 35% w/w aq. soln., Reagent Grade, Thermo Scientific Chemicals
Description
Tetraethylammonium hydroxide is a strong organic base and finds applications in photovoltaic chemicals, advanced ceramics, flat panel display chemicals, selective etching and transesterification catalysts.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Density | 1.023 g/mL |
Boiling Point | (decomposition) |
Odor | Amine-like |
Linear Formula | (CH3CH2)4NOH |
Refractive Index | 1.404 |
Quantity | 25 g |
UN Number | UN3267 |
Beilstein | 3912961 |
Sensitivity | Air Sensitive |
Merck Index | 14,9201 |
Show More |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.