missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetraethylammonium perchlorate, 98% (dry wt.), cont. ca 10% water, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
Description
Tetraethylammonium perchlorate is used as supporting electrolyte in polarographic measurements.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | >300°C |
Odor | Odorless |
Quantity | 50 g |
UN Number | UN1479 |
Beilstein | 3638093 |
Solubility Information | Soluble in water,acetone,acetonitrile and dimethyl sulfoxide. |
Formula Weight | 229.71 |
Percent Purity | 98% |
Concentration or Composition (by Analyte or Components) | Contains ≈ 10% water |
Physical Form | Wet Crystals |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'type03FormHeader'
missing translation for 'productTitle'