missing translation for 'onlineSavingsMsg'
Learn More

Tetraethylthiuram disulfide, 97%, Thermo Scientific Chemicals

Product Code. 11475576
Click to view available options
Quantity:
100 g
250 g
1000 g
5000 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Dopamine beta-hydroxylase inhibitor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97-77-8
Molecular Formula C10H20N2S4
Molecular Weight (g/mol) 296.524
MDL Number MFCD00009048
InChI Key AUZONCFQVSMFAP-UHFFFAOYSA-N
Synonym disulfiram, tetraethylthiuram disulfide, antabuse, alcophobin, antabus, anticol, bis diethylthiocarbamoyl disulfide, esperal, teturam, tetd
PubChem CID 3117
ChEBI CHEBI:4659
IUPAC Name diethylcarbamothioylsulfanyl N,N-diethylcarbamodithioate
SMILES CCN(CC)C(=S)SSC(=S)N(CC)CC

Specifications

Melting Point 69°C to 72°C
Density 1.3
Boiling Point 117°C (17 mmHg)
Quantity 250 g
UN Number UN3077
Beilstein 1712560
Merck Index 14,3364
Formula Weight 296.54
Percent Purity 97%
Chemical Name or Material Tetraethylthiuram disulfide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.