missing translation for 'onlineSavingsMsg'
Learn More

Tetraethylthiuram disulfide, 97%, Thermo Scientific Chemicals

Product Code. 10494545
Click to view available options
Quantity:
25 g
500 g
2.5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97-77-8
Molecular Formula C10H20N2S4
Molecular Weight (g/mol) 296.51
MDL Number MFCD00008157
InChI Key AUZONCFQVSMFAP-UHFFFAOYSA-N
Synonym disulfiram, tetraethylthiuram disulfide, antabuse, alcophobin, antabus, anticol, bis diethylthiocarbamoyl disulfide, esperal, teturam, tetd
PubChem CID 3117
ChEBI CHEBI:4659
IUPAC Name diethylcarbamothioylsulfanyl N,N-diethylcarbamodithioate
SMILES CCN(CC)C(=S)SSC(=S)N(CC)CC

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 70.0°C to 74.0°C
Color Yellow
Boiling Point 117°C (23.0 hPa)
Loss on Drying 0.3% max. (60°C)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Plastic bottle
Linear Formula [(C2H5)2NCS2-]2
Quantity 2.5 kg
Beilstein 04, 122
Fieser 06,569
Merck Index 15, 3407
Solubility Information Solubility in water: 0.02g/100mL. Other solubilities: 3.82g/100mL alcohol, 7.14g/100mL ether, soluble in acetone, benzene, carbon disulfice and, chloroform
Formula Weight 296.51
Percent Purity 97%
Physical Form Crystalline Powder, Crystals or Granules
Chemical Name or Material Tetraethylthiuram disulfide, 95%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Tetraethylthiuram disulfide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Respiratory sensitiser Category 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H373-May cause damage to organs.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.