missing translation for 'onlineSavingsMsg'
Learn More

Tetraethyltin, 98%, Thermo Scientific Chemicals

40.20 € - 563.81 €

Chemical Identifiers

CAS 597-64-8
Molecular Formula C8H20Sn
Molecular Weight (g/mol) 234.96
MDL Number MFCD00009020
InChI Key RWWNQEOPUOCKGR-UHFFFAOYSA-N
Synonym tetraethyltin, stannane, tetraethyl, tetraethyl tin, tin, tetraethyl, unii-31re6na75o, et4sn, snet4, c2h5 4sn, rwwnqeopuockgr-uhfffaoysa
PubChem CID 11704
IUPAC Name tetraethylstannane
SMILES CC[Sn](CC)(CC)CC
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11304839
View Documents
Thermo Scientific Alfa Aesar
071144.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11314839
View Documents
Thermo Scientific Alfa Aesar
071144.04
2 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11394829
View Documents
Thermo Scientific Alfa Aesar
071144.09
10 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11324839
View Documents
Thermo Scientific Alfa Aesar
071144.18
50 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Tetraethyltin is used in the electronics industry. Used as catalyst for olefin polymerization and to produce other organotin compounds. Also used as a preservative for wood, textiles, paper, and leather.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

597-64-8
234.96
RWWNQEOPUOCKGR-UHFFFAOYSA-N
11704
CC[Sn](CC)(CC)CC
C8H20Sn
MFCD00009020
tetraethyltin, stannane, tetraethyl, tetraethyl tin, tin, tetraethyl, unii-31re6na75o, et4sn, snet4, c2h5 4sn, rwwnqeopuockgr-uhfffaoysa
tetraethylstannane

Specifications

597-64-8
1.187g/mL
53°C (128°F)
C8H20Sn
MFCD00009020
tetraethyltin, stannane, tetraethyl, tetraethyl tin, tin, tetraethyl, unii-31re6na75o, et4sn, snet4, c2h5 4sn, rwwnqeopuockgr-uhfffaoysa
RWWNQEOPUOCKGR-UHFFFAOYSA-N
tetraethylstannane
11704
98%
Tetraethyltin
-112°C
181°C
Characteristic
1.4730
UN3384
Insoluble in water; Soluble in alcohol and ether
CC[Sn](CC)(CC)CC
234.96
234.94
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H300-H310-H330-H226
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Flammable liquid and vapor.

GHS P Statement
P273-P303+P361+P353-P309-P310-P501a
Avoid release to the environment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: TOXIC BY INHALATION LIQUID, FLAMMABLE, N.O.S.

missing translation for 'rtecsNumber' : WH8625000

missing translation for 'tsca' : TSCA

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only