missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrafluoroboric acid, ca 50% w/w aq. soln., Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
2500 g
Description
Tetrafluoroboric acid is used in conjunction with various methods of cleaning and to treat the surface of substrates. It is also used as a radiotracer.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Tetrafluoroboric acid is used in conjunction with various methods of cleaning and to treat the surface of substrates. It is also used as a radiotracer.
Solubility
Miscible with water.
Notes
Incompatible with strong bases, oxidizing agents and acids.
Tetrafluoroboric acid is used in conjunction with various methods of cleaning and to treat the surface of substrates. It is also used as a radiotracer.
Solubility
Miscible with water.
Notes
Incompatible with strong bases, oxidizing agents and acids.
Specifications
Specifications
Boiling Point | 130°C (decomposition) |
Concentration | ≈50% w/w aq. soln. |
Density | 1.4 |
Odor | Pungent |
Quantity | 500 g |
Solubility Information | Miscible with water. |
Formula Weight | 87.81 |
Concentration or Composition (by Analyte or Components) | ≈50% w/w aq. soln. |
Chemical Name or Material | Tetrafluoroboric acid |
Safety and Handling
Product Identifier
- Tetrafluoroboric acid, ca 50% w/w aqueous solution
- Danger
- Serious eye damage/eye irritation Category 1
- Reproductive toxicity Category 1B
- Skin corrosion/irritation Category 1 B
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : fluoroboric acid, boric acid
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction