Learn More
Tetrafluoroboric acid, ca 50% w/w aq. soln., Thermo Scientific Chemicals
24.60 € - 139.00 €
Chemical Identifiers
CAS | 16872-11-0 |
---|---|
MDL Number | MFCD00011345 |
Synonym | fluoroboric acid, fluoboric acid, tetrafluoroboric acid, hydrogen tetrafluoroborate, borofluoric acid, hydrofluoboric acid, borate 1-, tetrafluoro-, hydrogen, hydron tetrafluoroborate, hydrogen tetrafluoroborate 1-, unii-h429wz9fbq |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11368915
|
Thermo Scientific Alfa Aesar
L14037.22 |
100 g |
24.60 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11378915
|
Thermo Scientific Alfa Aesar
L14037.36 |
500 g |
38.40 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11388915
|
Thermo Scientific Alfa Aesar
L14037.0E |
2500 g |
139.00 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Tetrafluoroboric acid is used in conjunction with various methods of cleaning and to treat the surface of substrates. It is also used as a radiotracer.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsTetrafluoroboric acid is used in conjunction with various methods of cleaning and to treat the surface of substrates. It is also used as a radiotracer.
Solubility
Miscible with water.
Notes
Incompatible with strong bases, oxidizing agents and acids.
Chemical Identifiers
16872-11-0 | |
MFCD00011345 | |
fluoroboric acid, fluoboric acid, tetrafluoroboric acid, hydrogen tetrafluoroborate, borofluoric acid, hydrofluoboric acid, borate 1-, tetrafluoro-, hydrogen, hydron tetrafluoroborate, hydrogen tetrafluoroborate 1-, unii-h429wz9fbq |
Specifications
16872-11-0 | |
≈50% w/w aq. soln. | |
Pungent | |
fluoroboric acid, fluoboric acid, tetrafluoroboric acid, hydrogen tetrafluoroborate, borofluoric acid, hydrofluoboric acid, borate 1-, tetrafluoro-, hydrogen, hydron tetrafluoroborate, hydrogen tetrafluoroborate 1-, unii-h429wz9fbq | |
87.81 | |
Tetrafluoroboric acid |
130°C (decomposition) | |
1.4 | |
MFCD00011345 | |
Miscible with water. | |
≈50% w/w aq. soln. |
Safety and Handling
P201-P202-P260-P264b-P271-P281-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H360FD
missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: II
missing translation for 'einecsNumber' : 240-898-3
missing translation for 'rtecsNumber' : ED2685000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Tetrafluoroboric acid, ca 50% w/w aqueous solution
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Reproductive toxicity Category 1B
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : fluoroboric acid, boric acid
RUO – Research Use Only