missing translation for 'onlineSavingsMsg'
Learn More

Tetrafluoroboric acid, 49.5-50.5%, Honeywell Fluka™

Product Code. 15650550 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
1 L
This item is not returnable. View return policy
This item is not returnable. View return policy

49.5-50.5%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

CAS 16872-11-0
Concentration 49.5 to 50.5%
Density 1.38g/mL at 20°C
Quantity 1 L
Molecular Formula BF4H
Linear Formula HBF4
MDL Number MFCD00011345
UN Number 1775
Synonym fluoroboric acid, fluoboric acid, tetrafluoroboric acid, hydrogen tetrafluoroborate, borofluoric acid, hydrofluoboric acid, borate 1-, tetrafluoro-, hydrogen, hydron tetrafluoroborate, hydrogen tetrafluoroborate 1-, unii-h429wz9fbq
InChI Key ODGCEQLVLXJUCC-UHFFFAOYSA-O
SMILES [H+].[B-](F)(F)(F)F
IUPAC Name hydron;tetrafluoroborate
Molecular Weight (g/mol) 87.812
PubChem CID 28118
ChEBI CHEBI:38902
Formula Weight 87.81g/mol
Chemical Name or Material Tetrafluoroboric Acid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.