missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydro-2H-pyran-4-carboxylic acid, 97+%, Thermo Scientific™

Product Code. 10371003
Click to view available options
Quantity:
250 mg
1 g
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 5337-03-1
Molecular Formula C6H10O3
Molecular Weight (g/mol) 130.14
InChI Key AVPKHOTUOHDTLW-UHFFFAOYSA-N
Synonym tetrahydro-2h-pyran-4-carboxylic acid, tetrahydropyran-4-yl-carboxylic acid, tetrahydropyran-4-carboxylic acid, tetrahydro-pyran-4-carboxylic acid, 2h-pyran-4-carboxylic acid, tetrahydro, 4-carboxytetrahydro-2h-pyran, terahydropyran-4-yl-carboxylic acid, 3,4,5,6-tetrahydro-2h-pyran-4-carboxylic acid, 2h-3,4,5,6-tetrahydropyran-4-carboxylic acid
PubChem CID 219302

Specifications

Assay Percent Range 97%
Quantity 250 mg
Formula Weight 130.14
Percent Purity 97%
Chemical Name or Material Tetrahydro-2H-pyran-4-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'