missing translation for 'onlineSavingsMsg'
Learn More

tetrahydro-2h-pyran-4-ylmethanol, 97%, Thermo Scientific™

Product Code. 10036062
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 14774-37-9
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00457804
InChI Key YSNVSVCWTBLLRW-UHFFFAOYSA-N
Synonym tetrahydropyran-4-methanol, tetrahydro-2h-pyran-4-yl methanol, 4-hydroxymethyl tetrahydropyran, oxan-4-yl methanol, tetrahydro-pyran-4-yl-methanol, tetrahydro-2h-pyran-4-ylmethanol, 2h-pyran-4-methanol, tetrahydro, 4-hydroxymethyltetrahydropyran, oxan-4-yl-methanol
PubChem CID 2773573
SMILES OCC1CCOCC1

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
IUPAC Name oxan-4-ylmethanol
Formula Weight 116.16
Percent Purity 97%
Chemical Name or Material Tetrahydro-2H-pyran-4-ylmethanol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'