missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrahydro-4H-pyran-4-one, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Color | Colorless to Yellow |
Density | 1.0800g/mL |
Boiling Point | 166.0°C |
Flash Point | 56°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 99% |
Packaging | Glass bottle |
Refractive Index | 1.4510 to 1.4530 |
Quantity | 25 g |
Beilstein | 17,I,131 |
Show More |
Safety and Handling
Product Identifier
- tetrahydro-4H-pyran-4-one
- Warning
- Flammable liquid Category 3
- H226-Flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'