missing translation for 'onlineSavingsMsg'
Learn More
Tetrahydrofuran, 99.8%, for HPLC, unstabilized, Thermo Scientific Chemicals Product Code.: 10412441

Tetrahydrofuran, 99.8%, for HPLC, unstabilized, Thermo Scientific Chemicals
GREENER_CHOICE

Product Code. 10412441
2.5 L
Click to view available options
Quantity:
500 mL
1 L
2.5 L
This item is not returnable. View return policy

Product Code. 10412441

Brand: Thermo Scientific Acros 268290025

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For HPLC

EcoSafPak™ is an environmentally friendly packaging system made of 100% recyclable material by a SFI certified supplier.

Greener Choice Claims
  • Environmental benefits include:
    • Recycled Content
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-99-9
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00005356
InChI Key WYURNTSHIVDZCO-UHFFFAOYSA-N
Synonym tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofuranoShow MoreShow Less
PubChem CID 8028
ChEBI CHEBI:26911
IUPAC Name oxolane
SMILES C1CCOC1

Specifications

Absorbance (1 cm cell vs water), 0.02 max. at 280nm, 0.04 max. at 270nm, 0.05 max. at 260nm, 0.1 max. at 250nm, 0.26 max. at 240nm, 0.5 max. at 230nm
Acidity 0.0002 meq/g max.
Alkalinity 0.0006 meq/g max.
Density 0.8800g/mL
Quantity 2.5 L
Packaging Glass Bottle
Physical Form Liquid
Assay Percent Range 99.8%
Residue after Evaporation 0.0002% max.
Vapor Pressure Vapor Pressure: 200mbar at 20°C
Beilstein 17, 10
Water 0.03% max.
Fieser 01,1140; 02,398; 03,278; 05,649; 06,570
Merck Index 15, 9356
Formula Weight 72.11
Percent Purity ≥99.8%
Grade HPLC
Chemical Name or Material Tetrahydrofuran
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2528343 Certificate TypeCertificate of Analysis Date25/02/2025 Catalog Number268290025,268299_10979_60KG,268290010,268291_10771_500KG,268295000
Lot Number2413149 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number268290025,268290010,268295000
Lot Number2402240 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number268290025,268290010,268295000
Lot Number2402113 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number268290025,268290010,268295000
Lot Number2497796 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number268290025,268290010,268295000
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Tetrahydrofuran
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H302-Harmful if swallowed.
  • H225-Highly flammable liquid and vapour.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contains : Tetrahydrofuran

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tetrahydrofuran, 99.8%, for HPLC, unstabilized, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.