missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrahydrofuran, 99.8%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
AcroSeal™ Glass Bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | -108.4°C |
Density | 0.8800g/mL |
Boiling Point | 66.0°C |
Quantity | 1 L |
Flash Point | −21°C |
Packaging | AcroSeal™ Glass Bottle |
Refractive Index | 1.407 |
Assay Percent Range | 99.8% |
Specific Gravity | 0.88 |
Viscosity | 0.55 mPa.s (20°C) |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0459675 | Certificate of Analysis | 25/01/2025 | 450530025,450530010 |
A0459679 | Certificate of Analysis | 25/01/2025 | 450530025,450530010 |
A0463888 | Certificate of Analysis | 17/06/2024 | 450530025,450530010 |
A0459675 | Certificate of Analysis | 18/04/2024 | 450530025,450530010 |
A0459679 | Certificate of Analysis | 25/01/2024 | 450530025,450530010 |
Safety and Handling



Product Identifier
- Tetrahydrofuran
- Danger
- Acute toxicity Category 4
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
- H319-Causes serious eye irritation.
- H351-Suspected of causing cancer.
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
- H302-Harmful if swallowed.
- H225-Highly flammable liquid and vapour.
- EUH019-May form explosive peroxides.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Tetrahydrofuran, 99.8%, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction